Assay Details
Gene Symbol: AFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169123.1 | NP_001162594.1 | 5 - 6 | 1640 | 83 | |
NM_002025.3 | NP_002016.2 | 5 - 6 | 1652 | 83 | ||
GenBank mRNA | BC132683.1 | - | 5 - 6 | 1212 | 40146705 | 83 |
U48436.1 | - | 5 - 6 | 1652 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
