Assay Details
Gene Symbol: AFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169122.1 | NP_001162593.1 | 5 - 6 | 1596 | 84 | |
NM_001170628.1 | NP_001164099.1 | 3 - 4 | 229 | 84 | ||
NM_002025.3 | NP_002016.2 | 6 - 7 | 1695 | 84 | ||
GenBank mRNA | AK301927.1 | - | 3 - 4 | 229 | 84 | |
L76569.1 | - | 5 - 6 | 1509 | 84 | ||
U48436.1 | - | 6 - 7 | 1695 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
