Assay Details
Gene Symbol: YEATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018023.4 | NP_060493.3 | 3 - 4 | 393 | 128 | |
XM_005247597.2 | XP_005247654.1 | 3 - 4 | 407 | 128 | ||
XM_005247598.4 | XP_005247655.1 | 3 - 4 | 315 | 128 | ||
XM_006713688.3 | XP_006713751.1 | 3 - 4 | 525 | 128 | ||
XM_011512965.1 | XP_011511267.1 | 3 - 4 | 407 | 128 | ||
XM_011512966.1 | XP_011511268.1 | 3 - 4 | 407 | 128 | ||
XM_017006809.1 | XP_016862298.1 | 3 - 4 | 524 | 128 | ||
XM_017006810.1 | XP_016862299.1 | 3 - 4 | 315 | 128 | ||
XM_017006811.1 | XP_016862300.1 | 2 - 3 | 177 | 128 | ||
XM_017006812.1 | XP_016862301.1 | 3 - 4 | 414 | 128 | ||
XM_017006813.1 | XP_016862302.1 | 3 - 4 | 395 | 128 | ||
XM_017006814.1 | XP_016862303.1 | 3 - 4 | 304 | 128 | ||
GenBank mRNA | AB033023.1 | - | 3 - 4 | 393 | 128 | |
AK023370.1 | - | 2 - 3 | 196 | 128 | ||
BC150273.1 | - | 3 - 4 | 393 | 9007245 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000986.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK023370.1, XM_017006811.1, XM_017006813.1, XM_017006814.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03016954.hg.1 | ENST00000305135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03016954.hg.1 | ENST00000421660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03016954.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03016954.hg.1 | TC03000986.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0300169710.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300169710.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300169710.hg.1 | TC0300009677.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300169710.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300169710.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000305135.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000432781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000468850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000472593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000481343.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | ENST00000497765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009677.hg.1 | NM_018023.4 | Refseq |
