Assay Details
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011509307.2 | XP_011507609.2 | 19 - 20 | 2546 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
