Assay Details
Gene Symbol: ERBB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042599.1 | NP_001036064.1 | 16 - 17 | 2041 | 61 | |
NM_005235.2 | NP_005226.1 | 16 - 17 | 2041 | 61 | ||
XM_006712364.2 | XP_006712427.1 | 17 - 18 | 2605 | 61 | ||
XM_017003577.1 | XP_016859066.1 | 18 - 19 | 2683 | 61 | ||
XM_017003578.1 | XP_016859067.1 | 17 - 18 | 2638 | 61 | ||
XM_017003579.1 | XP_016859068.1 | 18 - 19 | 2683 | 61 | ||
XM_017003581.1 | XP_016859070.1 | 17 - 18 | 2638 | 61 | ||
XM_017003582.1 | XP_016859071.1 | 13 - 14 | 1718 | 61 | ||
GenBank mRNA | AB209697.1 | - | 6 - 7 | 745 | 61 | |
AK308606.1 | - | 16 - 17 | 1997 | 61 | ||
BC112199.1 | - | 16 - 17 | 1982 | 8327667 | 61 | |
BC143741.1 | - | 16 - 17 | 1970 | 9052255 | 61 | |
KT310076.1 | - | 5 - 6 | 728 | 61 | ||
L07868.1 | - | 16 - 17 | 1976 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002735.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
