Assay Details
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018448.4 | NP_060918.2 | 1 - 2 | 509 | 96 | |
GenBank mRNA | AB020636.2 | - | 1 - 2 | 411 | 96 | |
AK096010.1 | - | 1 - 2 | 415 | 96 | ||
AK292456.1 | - | 3 - 4 | 348 | 96 | ||
AK314358.1 | - | 1 - 2 | 397 | 96 | ||
AL136810.1 | - | 1 - 2 | 421 | 96 | ||
BC050341.1 | - | 1 - 2 | 205 | 5265409 | 96 | |
DB098973.1 | - | 1 - 2 | 486 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000595.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12007828.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007828.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007828.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007828.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007834.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007834.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007834.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007834.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007835.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007835.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007835.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007835.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007829.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007829.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007829.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007827.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007827.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007827.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007833.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007833.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007833.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007833.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12007832.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007832.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007832.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200200834.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200834.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200834.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200841.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200836.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200836.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200836.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200836.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200836.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200835.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200835.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200835.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200835.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200835.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200838.hg.1 | NM_018448.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 120765 | - | - |
