Assay Details
Gene Symbol: CAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018448.4 | NP_060918.2 | 2 - 3 | 653 | 161 | |
GenBank mRNA | AB020636.2 | - | 2 - 3 | 555 | 161 | |
AK096010.1 | - | 2 - 3 | 559 | 161 | ||
AK292456.1 | - | 4 - 5 | 492 | 161 | ||
AK314358.1 | - | 2 - 3 | 541 | 161 | ||
AL136810.1 | - | 2 - 3 | 565 | 161 | ||
BC050341.1 | - | 2 - 3 | 349 | 5265409 | 161 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000595.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12007836.hg.1 | ENST00000299218.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007836.hg.1 | ENST00000545606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12007836.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12007836.hg.1 | TC12000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200844.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200200844.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200844.hg.1 | TC1200012657.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200200844.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200200844.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000535146.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539109.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000539434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000540525.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000541058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | ENST00000544619.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012657.hg.1 | NM_018448.4 | Refseq |
