Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004943.1 | NP_004934.1 | 1 - 2 | 484 | 85 | |
GenBank mRNA | BM922103.1 | - | 484 | 85 | ||
L19267.1 | - | 1 - 2 | 72 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001633.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19022314.hg.1 | TC19001633.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19022314.hg.1 | ENST00000270223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19022314.hg.1 | ENST00000377735.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19022314.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19022315.hg.1 | TC19001633.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19022315.hg.1 | ENST00000270223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19022315.hg.1 | ENST00000377735.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19022315.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19022316.hg.1 | TC19001633.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19022316.hg.1 | ENST00000377735.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19022316.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | TC1900011985.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206898.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | TC1900011985.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206899.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | TC1900011985.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206901.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | TC1900011985.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206897.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | TC1900011985.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206896.hg.1 | NM_004943.1 | Refseq |
