Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC019266.1 | - | 1 - 2 | 1030 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000270223.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000377735.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000537879.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000597053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000598237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000601370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | ENST00000602829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011985.hg.1 | NM_004943.1 | Refseq |
