Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000287380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000309336.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000327098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000378080.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000518805.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000521676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
ENST00000522420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
NM_001145088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
NM_145647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009640.hg.1 |
TC08000712.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000287380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000327098.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000518577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000518805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000521676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000522420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000524307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
NM_001145088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
NM_145647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
TC0800008726.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000287380.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000327098.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000518577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000518805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000521676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000522420.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
ENST00000524307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
NM_001145088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159777.hg.1 |
NM_145647.3 |
Refseq |