Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK000839.1 | - | 1 | 1890 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 10a | MIR10A |
MIRN10A, hsa-mir-10a, miRNA10A, mir-10a |
-370.0 | Sense | Chr. 17 - 48579838 - 48579947 on NCBI Build GRCh38 |
Downstream | HOXB cluster antisense RNA 3 | HOXB-AS3 | 8.2kb | Anti-sense | Chr. 17 - 48590420 - 48606414 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
