Assay Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004052.3 | NP_004043.3 | 1 - 2 | 253 | 85 | |
GenBank mRNA | AF002697.1 | - | 1 - 2 | 172 | 85 | |
AK222626.1 | - | 1 - 2 | 163 | 85 | ||
AK295136.1 | - | 1 - 2 | 163 | 85 | ||
BC009342.2 | - | 1 - 2 | 148 | 85 | ||
BC021989.1 | - | 1 - 2 | 167 | 4066029 | 85 | |
BC067818.1 | - | 1 - 2 | 196 | 85 | ||
BC080643.1 | - | 1 - 2 | 148 | 85 | ||
HY049371.1 | - | 1 - 2 | 254 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001769.hg
Additional Information:
This assay is in the 5'UTR region of the AF002697.1,AK295136.1,BC021989.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy
response to hypoxia
apoptotic process
cell death
granzyme-mediated apoptotic signaling pathway
positive regulation of autophagy
negative regulation of mitochondrial fusion
positive regulation of cardiac muscle cell apoptotic process
positive regulation of macroautophagy
mitochondrial protein catabolic process
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of protein complex disassembly
mitochondrial fragmentation involved in apoptotic process
negative regulation of membrane potential
regulation of mitochondrial membrane permeability
autophagic cell death
brown fat cell differentiation
neuron apoptotic process
defense response to virus
response to hyperoxia
negative regulation of cell death
cellular response to hydrogen peroxide
cellular response to mechanical stimulus
cellular response to cobalt ion
cellular response to hypoxia
reactive oxygen species metabolic process
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
mitochondrial outer membrane permeabilization
toxin transport
regulation of aerobic respiration
intrinsic apoptotic signaling pathway in response to hypoxia
negative regulation of reactive oxygen species metabolic process
protein binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
GTPase binding
mitophagy
response to hypoxia
apoptotic process
cell death
granzyme-mediated apoptotic signaling pathway
positive regulation of autophagy
negative regulation of mitochondrial fusion
positive regulation of cardiac muscle cell apoptotic process
positive regulation of macroautophagy
mitochondrial protein catabolic process
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of protein complex disassembly
mitochondrial fragmentation involved in apoptotic process
negative regulation of membrane potential
regulation of mitochondrial membrane permeability
autophagic cell death
brown fat cell differentiation
neuron apoptotic process
defense response to virus
response to hyperoxia
negative regulation of cell death
cellular response to hydrogen peroxide
cellular response to mechanical stimulus
cellular response to cobalt ion
cellular response to hypoxia
reactive oxygen species metabolic process
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
mitochondrial outer membrane permeabilization
toxin transport
regulation of aerobic respiration
intrinsic apoptotic signaling pathway in response to hypoxia
negative regulation of reactive oxygen species metabolic process
protein binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
GTPase binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010085.hg.1 | ENST00000621794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010085.hg.1 | ENST00000621794.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023346.hg.1 | ENST00000368636.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023346.hg.1 | ENST00000540159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023346.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023346.hg.1 | TC10001769.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900010105.hg.1 | ENST00000605397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010111.hg.1 | ENST00000599655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007455.hg.1 | ENST00000603714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007463.hg.1 | ENST00000604600.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006783.hg.1 | ENST00000547295.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007576.hg.1 | ENST00000600724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023345.hg.1 | ENST00000368636.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023345.hg.1 | ENST00000540159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023345.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023345.hg.1 | TC10001769.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007549.hg.1 | ENST00000601184.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007549.hg.1 | ENST00000601184.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184069.hg.1 | TC1000012329.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184070.hg.1 | TC1000012329.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900010085.hg.1 | ENST00000621794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010085.hg.1 | ENST00000621794.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010105.hg.1 | ENST00000605397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010111.hg.1 | ENST00000599655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184071.hg.1 | TC1000012329.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000540159.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012329.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007463.hg.1 | ENST00000604600.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000631806.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | ENST00000633835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184073.hg.1 | TC1000012329.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900007549.hg.1 | ENST00000601184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007549.hg.1 | ENST00000601184.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006783.hg.1 | ENST00000547295.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006783.hg.1 | ENST00000550043.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184075.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184075.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184075.hg.1 | ENST00000368636.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184075.hg.1 | NM_004052.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184075.hg.1 | TC1000012329.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900007455.hg.1 | ENST00000603714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007576.hg.1 | ENST00000600724.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 127211 | - | - |
