Assay Details
Gene Symbol: MAPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001292039.1 | NP_001278968.1 | 2 - 3 | 279 | 81 | |
NM_002747.3 | NP_002738.2 | 3 - 4 | 1695 | 81 | ||
XM_005258299.3 | XP_005258356.1 | 3 - 4 | 6057 | 81 | ||
XM_011526074.2 | XP_011524376.1 | 3 - 4 | 6239 | 81 | ||
XM_011526075.2 | XP_011524377.1 | 3 - 4 | 1697 | 81 | ||
XM_011526076.2 | XP_011524378.1 | 3 - 4 | 2060 | 81 | ||
XM_011526077.2 | XP_011524379.1 | 2 - 3 | 2706 | 81 | ||
XM_017025839.1 | XP_016881328.1 | 3 - 4 | 1628 | 81 | ||
GenBank mRNA | AK293818.1 | - | 2 - 3 | 260 | 81 | |
AK295058.1 | - | 3 - 4 | 1616 | 81 | ||
AK303601.1 | - | 2 - 3 | 357 | 81 | ||
BC117216.1 | - | 2 - 3 | 749 | 40125767 | 81 | |
X59727.1 | - | 2 - 3 | 1232 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000179.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18002084.hg.1 | ENST00000400384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002084.hg.1 | ENST00000540640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002084.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002084.hg.1 | TC18000179.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000586735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000586735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | TC1800007320.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150071.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000586735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000586735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000587823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000588916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007320.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000400384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | TC1800007320.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000592595.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_001292039.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_001292040.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | NM_002747.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000540640.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150057.hg.1 | ENST00000400384.6 | Ensembl |
