Assay Details
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014423.3 | NP_055238.1 | 15 - 16 | 3204 | 144 | |
XM_005271963.4 | XP_005272020.1 | 16 - 17 | 3268 | 144 | ||
XM_005271964.4 | XP_005272021.1 | 9 - 10 | 1916 | 144 | ||
XM_006714587.3 | XP_006714650.1 | 14 - 15 | 3117 | 144 | ||
GenBank mRNA | AF197927.1 | - | 15 - 16 | 3133 | 144 | |
AF213987.2 | - | 15 - 16 | 2861 | 144 | ||
BC137226.1 | - | 15 - 16 | 3102 | 9021223 | 144 | |
BC144538.1 | - | 16 - 17 | 3119 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001776.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
