Assay Details
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014423.3 | NP_055238.1 | 19 - 20 | 3553 | 109 | |
XM_005271963.4 | XP_005272020.1 | 20 - 21 | 3617 | 109 | ||
XM_005271964.4 | XP_005272021.1 | 13 - 14 | 2265 | 109 | ||
XM_006714587.3 | XP_006714650.1 | 18 - 19 | 3466 | 109 | ||
GenBank mRNA | AF197927.1 | - | 19 - 20 | 3482 | 109 | |
AF213987.2 | - | 19 - 20 | 3210 | 109 | ||
BC137226.1 | - | 19 - 20 | 3451 | 9021223 | 109 | |
BC144538.1 | - | 20 - 21 | 3468 | 109 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001776.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
