Assay Details
Gene Symbol: DESI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016076.4 | NP_057160.2 | 3 - 4 | 473 | 73 | |
XM_011544203.2 | XP_011542505.1 | 4 - 5 | 558 | 73 | ||
XM_011544205.2 | XP_011542507.1 | 3 - 4 | 1124 | 73 | ||
GenBank mRNA | AF151904.1 | - | 3 - 4 | 265 | 73 | |
AK314664.1 | - | 3 - 4 | 443 | 73 | ||
BC004485.2 | - | 3 - 4 | 420 | 3831754 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001989.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030830.hg.1 | ENST00000263831.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030830.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030830.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030830.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030831.hg.1 | ENST00000263831.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030831.hg.1 | ENST00000302550.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030831.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030831.hg.1 | TC01001989.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190009.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | ENST00000484738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012222.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NR_123736.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | TC0100012222.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000263831.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000302550.15 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | ENST00000418162.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NM_001297746.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NM_016076.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NR_123735.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190008.hg.1 | NR_123736.1 | Refseq |
