Assay Details
Gene Symbol: CHRNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000080.3 | NP_000071.1 | 9 - 10 | 1042 | 144 | |
XM_017024115.1 | XP_016879604.1 | 9 - 10 | 1061 | 144 | ||
GenBank mRNA | X66403.1 | - | 9 - 10 | 1042 | 144 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001032.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
transport
muscle contraction
signal transduction
synaptic transmission, cholinergic
neuromuscular synaptic transmission
response to nicotine
regulation of membrane potential
cation transmembrane transport
cation transmembrane transporter activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
skeletal muscle contraction
transport
muscle contraction
signal transduction
synaptic transmission, cholinergic
neuromuscular synaptic transmission
response to nicotine
regulation of membrane potential
cation transmembrane transport
cation transmembrane transporter activity
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17013904.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013904.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013904.hg.1 | TC17001032.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | ENST00000575637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009535.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | ENST00000572438.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | TC1700009535.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | NM_000080.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | ENST00000293780.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700173375.hg.1 | ENST00000572438.1 | Ensembl |
