Assay Details

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000442.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
