Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014051.hg.1 |
TC22000829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014051.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014051.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014051.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014052.hg.1 |
TC22000829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014052.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014052.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014052.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000468151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008832.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
TC2200008832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164015.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
NM_015704.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
TC2200008832.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000263256.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000463886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
ENST00000482612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164016.hg.1 |
NM_015704.2 |
Refseq |