Assay Details
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004947.4 | NP_004938.1 | 31 - 32 | 3274 | 96 | |
XM_005264914.3 | XP_005264971.1 | 31 - 32 | 3546 | 96 | ||
XM_005264915.3 | XP_005264972.1 | 31 - 32 | 3546 | 96 | ||
XM_005264916.3 | XP_005264973.1 | 31 - 32 | 3324 | 96 | ||
XM_005264917.3 | XP_005264974.1 | 31 - 32 | 3504 | 96 | ||
XM_005264918.3 | XP_005264975.1 | 30 - 31 | 3176 | 96 | ||
XM_006713008.3 | XP_006713071.1 | 31 - 32 | 3546 | 96 | ||
XM_006713009.3 | XP_006713072.1 | 31 - 32 | 3546 | 96 | ||
XM_006713010.3 | XP_006713073.1 | 31 - 32 | 3309 | 96 | ||
XM_011533441.2 | XP_011531743.1 | 20 - 21 | 4155 | 96 | ||
XM_011533443.2 | XP_011531745.1 | 7 - 8 | 1836 | 96 | ||
XM_011533444.2 | XP_011531746.1 | 7 - 8 | 1760 | 96 | ||
XM_011533445.2 | XP_011531747.1 | 7 - 8 | 1781 | 96 | ||
XM_017005825.1 | XP_016861314.1 | 31 - 32 | 3325 | 96 | ||
XM_017005826.1 | XP_016861315.1 | 31 - 32 | 3315 | 96 | ||
XM_017005827.1 | XP_016861316.1 | 22 - 23 | 2466 | 96 | ||
XM_017005828.1 | XP_016861317.1 | 6 - 7 | 1191 | 96 | ||
GenBank mRNA | AB002297.1 | - | 26 - 27 | 2882 | 96 | |
AY145303.1 | - | 31 - 32 | 3251 | 96 | ||
AY254099.1 | - | 31 - 32 | 3274 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000318.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
