Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016701.hg.1 |
ENST00000358514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016701.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016701.hg.1 |
TC16002086.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016700.hg.1 |
ENST00000358514.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016700.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16016700.hg.1 |
TC16002086.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011553.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
TC1600011553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194020.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
NM_002801.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
TC1600011553.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000358514.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000570304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000570985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000574576.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
ENST00000575556.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194017.hg.1 |
NM_002801.3 |
Refseq |