Assay Details
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286810.1 | NP_001273739.1 | 4 - 5 | 608 | 89 | |
NM_024345.4 | NP_077321.3 | 4 - 5 | 1134 | 89 | ||
XM_005251578.3 | XP_005251635.1 | 3 - 4 | 957 | 89 | ||
XM_017015129.1 | XP_016870618.1 | 3 - 4 | 444 | 89 | ||
GenBank mRNA | AK026854.1 | - | 2 - 3 | 275 | 89 | |
AK074532.1 | - | 4 - 5 | 621 | 89 | ||
AK309712.1 | - | 4 - 5 | 608 | 89 | ||
AK315756.1 | - | 4 - 5 | 587 | 89 | ||
BC011959.1 | - | 2 - 3 | 337 | 89 | ||
BC108686.1 | - | 4 - 5 | 1103 | 6157119 | 89 | |
BC139834.1 | - | 4 - 5 | 621 | 89 | ||
CD299973.1 | - | 4 - 5 | 649 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000218.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09002595.hg.1 | ENST00000377724.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002595.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002595.hg.1 | TC09000218.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | TC0900007167.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148915.hg.1 | NM_024345.4 | Refseq |
