Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006325.hg.1 |
ENST00000262630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006325.hg.1 |
ENST00000354109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006325.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006325.hg.1 |
TC19000481.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006319.hg.1 |
ENST00000262630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006319.hg.1 |
ENST00000354109.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006319.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006319.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006319.hg.1 |
TC19000481.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
TC1900007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158463.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
TC1900007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158457.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |