Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003587.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003587.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003587.hg.1 |
TC04000245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003585.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003585.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003585.hg.1 |
TC04000245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003586.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003586.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003586.hg.1 |
TC04000245.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000509518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
ENST00000509518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150059.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150059.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150059.hg.1 |
TC0400007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150059.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150059.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150061.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150061.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150061.hg.1 |
TC0400007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150061.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150061.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150058.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150058.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150058.hg.1 |
TC0400007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150058.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150058.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150060.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150060.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150060.hg.1 |
TC0400007315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150060.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150060.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000310169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000502377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000509518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
ENST00000509518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007315.hg.1 |
NM_017581.3 |
Refseq |