Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003929.hg.1 |
ENST00000295454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003929.hg.1 |
ENST00000538619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003929.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003929.hg.1 |
TC04000273.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000381582.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000509366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000510909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
ENST00000513567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007419.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150667.hg.1 |
ENST00000295454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150667.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150667.hg.1 |
TC0400007419.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400150667.hg.1 |
NM_000812.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150667.hg.1 |
ENST00000295454.7 |
Ensembl |