Assay Details
Gene Symbol: LOC730102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BM127544.1 | - | 3 - 4 | 239 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000462729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000476232.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000481349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | ENST00000512906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018538.hg.1 | NR_037167.1 | Refseq |
