Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
ENST00000261892.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
ENST00000339868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
ENST00000399033.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
ENST00000537259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
ENST00000546330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
NM_004727.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004946.hg.1 |
TC15000607.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000261892.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000339868.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000399033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000434116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000537259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000546330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_004727.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
TC1500007602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000261892.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000339868.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000399033.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000434116.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000537259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
ENST00000546330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_001301033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153830.hg.1 |
NM_004727.2 |
Refseq |