Assay Details
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118886.1 | NP_001112358.1 | 3 - 4 | 1032 | 79 | |
XM_017029427.1 | XP_016884916.1 | 4 - 5 | 576 | 79 | ||
XM_017029428.1 | XP_016884917.1 | 4 - 5 | 450 | 79 | ||
XM_017029429.1 | XP_016884918.1 | 5 - 6 | 995 | 79 | ||
GenBank mRNA | AK289561.1 | - | 3 - 4 | 559 | 79 | |
AY437084.1 | - | 3 - 4 | 272 | 79 | ||
CR450343.1 | - | 3 - 4 | 272 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017029428.1, XM_017029429.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
