Assay Details
Gene Symbol: BMPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004329.2 | NP_004320.2 | 2 - 3 | 393 | 97 | |
XM_011540103.2 | XP_011538405.1 | 2 - 3 | 290 | 97 | ||
GenBank mRNA | AK291764.1 | - | 2 - 3 | 262 | 97 | |
BC028383.1 | - | 2 - 3 | 387 | 4824378 | 97 | |
Z22535.1 | - | 2 - 3 | 154 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000609.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291764.1, BC028383.1, NM_004329.2, XM_011540103.2, Z22535.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mesoderm formation
somitogenesis
Mullerian duct regression
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
outflow tract septum morphogenesis
outflow tract morphogenesis
cardiac conduction system development
mitral valve morphogenesis
tricuspid valve morphogenesis
endocardial cushion morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular compact myocardium morphogenesis
endocardial cushion formation
protein phosphorylation
immune response
transforming growth factor beta receptor signaling pathway
ectoderm development
dorsal/ventral axis specification
regulation of cardiac muscle cell apoptotic process
positive regulation of pathway-restricted SMAD protein phosphorylation
neural crest cell development
negative regulation of smooth muscle cell migration
stem cell population maintenance
pituitary gland development
neural plate mediolateral regionalization
lung development
positive regulation of bone mineralization
BMP signaling pathway
hindlimb morphogenesis
dorsal aorta morphogenesis
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
positive regulation of osteoblast differentiation
positive regulation of transcription by RNA polymerase II
paraxial mesoderm structural organization
lateral mesoderm development
regulation of lateral mesodermal cell fate specification
mesendoderm development
embryonic organ development
developmental growth
positive regulation of epithelial cell proliferation
negative regulation of neurogenesis
roof of mouth development
regulation of cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
positive regulation of SMAD protein signal transduction
heart formation
pharyngeal arch artery morphogenesis
cellular response to BMP stimulus
positive regulation of cardiac ventricle development
positive regulation of vascular associated smooth muscle cell proliferation
regulation of cellular senescence
DNA-binding transcription factor activity, RNA polymerase II-specific
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
protein binding
ATP binding
protein homodimerization activity
SMAD binding
metal ion binding
BMP receptor activity
in utero embryonic development
mesoderm formation
somitogenesis
Mullerian duct regression
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
outflow tract septum morphogenesis
outflow tract morphogenesis
cardiac conduction system development
mitral valve morphogenesis
tricuspid valve morphogenesis
endocardial cushion morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular compact myocardium morphogenesis
endocardial cushion formation
protein phosphorylation
immune response
transforming growth factor beta receptor signaling pathway
ectoderm development
dorsal/ventral axis specification
regulation of cardiac muscle cell apoptotic process
positive regulation of pathway-restricted SMAD protein phosphorylation
neural crest cell development
negative regulation of smooth muscle cell migration
stem cell population maintenance
pituitary gland development
neural plate mediolateral regionalization
lung development
positive regulation of bone mineralization
BMP signaling pathway
hindlimb morphogenesis
dorsal aorta morphogenesis
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
positive regulation of osteoblast differentiation
positive regulation of transcription by RNA polymerase II
paraxial mesoderm structural organization
lateral mesoderm development
regulation of lateral mesodermal cell fate specification
mesendoderm development
embryonic organ development
developmental growth
positive regulation of epithelial cell proliferation
negative regulation of neurogenesis
roof of mouth development
regulation of cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
positive regulation of SMAD protein signal transduction
heart formation
pharyngeal arch artery morphogenesis
cellular response to BMP stimulus
positive regulation of cardiac ventricle development
positive regulation of vascular associated smooth muscle cell proliferation
regulation of cellular senescence
DNA-binding transcription factor activity, RNA polymerase II-specific
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
protein binding
ATP binding
protein homodimerization activity
SMAD binding
metal ion binding
BMP receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10006638.hg.1 | ENST00000224764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006638.hg.1 | ENST00000372037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006638.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006638.hg.1 | TC10000609.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006639.hg.1 | ENST00000224764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006639.hg.1 | ENST00000372037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006639.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006639.hg.1 | TC10000609.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10006640.hg.1 | ENST00000224764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006640.hg.1 | ENST00000372037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006640.hg.1 | ENST00000480152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006640.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006640.hg.1 | TC10000609.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10006643.hg.1 | ENST00000224764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006643.hg.1 | ENST00000372037.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006643.hg.1 | ENST00000480152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006643.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006643.hg.1 | TC10000609.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000156342.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156342.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156342.hg.1 | TC1000008315.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156342.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156342.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156343.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156343.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156343.hg.1 | TC1000008315.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156343.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156343.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | TC1000008315.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156344.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | TC1000008315.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156346.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000372037.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | ENST00000480152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008315.hg.1 | NM_004329.2 | Refseq |
