Assay Details
Gene Symbol: GBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199378.1 | NP_001186307.1 | 28 - 29 | 3802 | 68 | |
NM_001199379.1 | NP_001186308.1 | 28 - 29 | 3799 | 68 | ||
NM_004193.2 | NP_004184.1 | 28 - 29 | 3799 | 68 | ||
XM_005270261.1 | XP_005270318.1 | 28 - 29 | 3802 | 68 | ||
XM_006718047.1 | XP_006718110.1 | 29 - 30 | 3901 | 68 | ||
XM_006718048.1 | XP_006718111.1 | 29 - 30 | 3898 | 68 | ||
XM_006718049.1 | XP_006718112.1 | 29 - 30 | 3901 | 68 | ||
XM_006718050.1 | XP_006718113.1 | 29 - 30 | 3865 | 68 | ||
XM_011540312.1 | XP_011538614.1 | 28 - 29 | 3760 | 68 | ||
XM_011540313.1 | XP_011538615.1 | 29 - 30 | 3901 | 68 | ||
XM_011540314.2 | XP_011538616.1 | 17 - 18 | 2279 | 68 | ||
XM_017016861.1 | XP_016872350.1 | 29 - 30 | 3877 | 68 | ||
XM_017016862.1 | XP_016872351.1 | 29 - 30 | 3865 | 68 | ||
XM_017016863.1 | XP_016872352.1 | 28 - 29 | 3763 | 68 | ||
XM_017016864.1 | XP_016872353.1 | 24 - 25 | 3101 | 68 | ||
GenBank mRNA | AF068755.1 | - | 28 - 29 | 3745 | 68 | |
AK024916.1 | - | 8 - 9 | 912 | 68 | ||
AK025153.1 | - | 1 - 2 | 63 | 68 | ||
AK025330.1 | - | 28 - 29 | 3768 | 68 | ||
BC117681.1 | - | 28 - 29 | 3626 | 40070852 | 68 | |
BC117682.1 | - | 28 - 29 | 3626 | 40070853 | 68 | |
D87435.2 | - | 28 - 29 | 3729 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000752.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
