Assay Details
Gene Symbol: GBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199378.1 | NP_001186307.1 | 37 - 38 | 5274 | 108 | |
NM_001199379.1 | NP_001186308.1 | 37 - 38 | 5271 | 108 | ||
NM_004193.2 | NP_004184.1 | 37 - 38 | 5283 | 108 | ||
XM_005270261.1 | XP_005270318.1 | 37 - 38 | 5286 | 108 | ||
XM_006718047.1 | XP_006718110.1 | 38 - 39 | 5385 | 108 | ||
XM_006718048.1 | XP_006718111.1 | 38 - 39 | 5382 | 108 | ||
XM_006718049.1 | XP_006718112.1 | 38 - 39 | 5373 | 108 | ||
XM_006718050.1 | XP_006718113.1 | 38 - 39 | 5349 | 108 | ||
XM_011540312.1 | XP_011538614.1 | 37 - 38 | 5244 | 108 | ||
XM_011540314.2 | XP_011538616.1 | 26 - 27 | 3763 | 108 | ||
XM_017016861.1 | XP_016872350.1 | 38 - 39 | 5361 | 108 | ||
XM_017016862.1 | XP_016872351.1 | 38 - 39 | 5337 | 108 | ||
XM_017016863.1 | XP_016872352.1 | 37 - 38 | 5247 | 108 | ||
XM_017016864.1 | XP_016872353.1 | 33 - 34 | 4585 | 108 | ||
GenBank mRNA | AF068755.1 | - | 37 - 38 | 5229 | 108 | |
AK024916.1 | - | 17 - 18 | 2384 | 108 | ||
AK025153.1 | - | 10 - 11 | 1547 | 108 | ||
AK025330.1 | - | 37 - 38 | 5240 | 108 | ||
BC007941.2 | - | 3 - 4 | 303 | 108 | ||
BC014171.2 | - | 7 - 8 | 1057 | 108 | ||
BC117681.1 | - | 37 - 38 | 5098 | 40070852 | 108 | |
BC117682.1 | - | 37 - 38 | 5098 | 40070853 | 108 | |
D87435.2 | - | 37 - 38 | 5213 | 108 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000752.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
