Assay Details
Gene Symbol: PAFAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000437.3 | NP_000428.2 | 3 - 4 | 426 | 143 | |
XM_006710670.3 | XP_006710733.1 | 3 - 4 | 491 | 143 | ||
XM_011541528.2 | XP_011539830.1 | 4 - 5 | 557 | 143 | ||
GenBank mRNA | AK313815.1 | - | 3 - 4 | 353 | 143 | |
BC001158.1 | - | 3 - 4 | 371 | 3354450 | 143 | |
D87845.1 | - | 3 - 4 | 449 | 143 | ||
U89386.1 | - | 3 - 4 | 325 | 143 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002384.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01036746.hg.1 | ENST00000374282.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036746.hg.1 | ENST00000374284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036746.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036746.hg.1 | TC01002384.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | NM_000437.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | TC0100013371.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000441420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000374282.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000374284.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | ENST00000439092.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199322.hg.1 | NM_000437.3 | Refseq |
