Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194463.1 | NP_919445.1 | 1 - 2 | 737 | 68 | |
GenBank mRNA | AF394689.1 | - | 1 - 2 | 497 | 68 | |
AK074264.1 | - | 1 - 2 | 737 | 68 | ||
BC063404.1 | - | 1 - 2 | 576 | 5191785 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000519.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X006927.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006927.hg.1 | ENST00000324342.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006927.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006927.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006927.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006925.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006925.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006925.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006926.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006926.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006926.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155057.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155054.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155055.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155055.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155055.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155055.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155055.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155056.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155056.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155056.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155056.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155056.hg.1 | NM_194463.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 160343 | - | - |
