Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024539.3 | NP_078815.3 | 2 - 3 | 821 | 81 | |
NM_194463.1 | NP_919445.1 | 2 - 3 | 984 | 81 | ||
GenBank mRNA | AK074264.1 | - | 2 - 3 | 984 | 81 | |
AK126553.1 | - | 2 - 3 | 804 | 81 | ||
BC030951.1 | - | 1 - 2 | 97 | 81 | ||
BC036901.1 | - | 2 - 3 | 833 | 81 | ||
BC063404.1 | - | 2 - 3 | 823 | 5191785 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000519.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X006923.hg.1 | ENST00000324342.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006923.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006923.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006922.hg.1 | ENST00000324342.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006922.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006922.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006928.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006928.hg.1 | ENST00000324342.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006928.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006928.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006928.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006924.hg.1 | ENST00000324342.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006924.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006924.hg.1 | TC0X000519.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0X00155051.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155051.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155051.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155051.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155051.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008054.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155050.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155052.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155052.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155052.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155052.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155052.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155053.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | NM_194463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | TC0X00008054.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000255499.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000324342.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | ENST00000418562.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | NM_024539.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155058.hg.1 | NM_194463.1 | Refseq |
