Assay Details
Gene Symbol: TDO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005651.3 | NP_005642.1 | 1 - 2 | 97 | 81 | |
GenBank mRNA | AK289418.1 | - | 1 - 2 | 94 | 81 | |
AK307741.1 | - | 1 - 2 | 94 | 81 | ||
BC005355.1 | - | 1 - 2 | 94 | 4071714 | 81 | |
BG616664.1 | - | 1 - 2 | 97 | 81 | ||
U32989.1 | - | 1 - 2 | 95 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000789.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04010895.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010895.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04010895.hg.1 | TC04000789.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04010894.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04010894.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04010894.hg.1 | TC04000789.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | TC0400009090.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162459.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000503634.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506072.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000506181.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000507590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000510293.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000512584.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009090.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | TC0400009090.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162458.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | NM_005651.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | TC0400009090.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | ENST00000509738.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | ENST00000536354.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400162457.hg.1 | NM_005651.3 | Refseq |
