Assay Details
Gene Symbol: AKAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004274.4 | NP_004265.3 | 2 - 3 | 493 | 85 | |
XM_017021808.1 | XP_016877297.1 | 1 - 2 | 357 | 85 | ||
GenBank mRNA | AB002309.2 | - | 2 - 3 | 433 | 85 | |
BC137232.1 | - | 2 - 3 | 376 | 9021229 | 85 | |
BC137233.1 | - | 2 - 3 | 376 | 9021230 | 85 | |
BC150185.1 | - | 2 - 3 | 422 | 85 | ||
BC150288.1 | - | 2 - 3 | 433 | 8860126 | 85 | |
BC154413.1 | - | 2 - 3 | 422 | 85 | ||
BP230135.1 | - | 2 - 3 | 493 | 85 | ||
U17195.2 | - | 2 - 3 | 449 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000208.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
