Assay Details
Gene Symbol: ADIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024551.2 | NP_078827.2 | 4 - 5 | 666 | 148 | |
XM_005253789.1 | XP_005253846.1 | 4 - 5 | 788 | 148 | ||
XM_006719018.2 | XP_006719081.1 | 5 - 6 | 785 | 148 | ||
XM_011521024.2 | XP_011519326.1 | 5 - 6 | 796 | 148 | ||
GenBank mRNA | AK127196.1 | - | 5 - 6 | 721 | 148 | |
AK128511.1 | - | 4 - 5 | 720 | 148 | ||
AY424280.1 | - | 3 - 4 | 545 | 148 | ||
BC051858.2 | - | 4 - 5 | 666 | 6527208 | 148 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000018.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G protein-coupled receptor signaling pathway
heart development
female pregnancy
response to nutrient
hormone-mediated signaling pathway
fatty acid oxidation
negative regulation of cell growth
adiponectin-activated signaling pathway
glucose homeostasis
positive regulation of glucose import
vascular wound healing
identical protein binding
metal ion binding
protein heterodimerization activity
adiponectin binding
adipokinetic hormone receptor activity
G protein-coupled receptor signaling pathway
heart development
female pregnancy
response to nutrient
hormone-mediated signaling pathway
fatty acid oxidation
negative regulation of cell growth
adiponectin-activated signaling pathway
glucose homeostasis
positive regulation of glucose import
vascular wound healing
identical protein binding
metal ion binding
protein heterodimerization activity
adiponectin binding
adipokinetic hormone receptor activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12000262.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000262.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000262.hg.1 | TC12000018.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000537545.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000540974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000543456.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | ENST00000544470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006472.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | TC1200006472.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000535774.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144457.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | NM_024551.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | TC1200006472.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | ENST00000357103.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | ENST00000537190.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144458.hg.1 | NM_024551.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 139014 | - | - |
