Assay Details
Gene Symbol: FAM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005264689.4 | XP_005264746.1 | 1 - 2 | 315 | 110 | |
XM_006711879.3 | XP_006711942.1 | 1 - 2 | 316 | 110 | ||
XM_011510342.2 | XP_011508644.1 | 1 - 2 | 327 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005264689.4, XM_006711879.3, XM_011510342.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of neuron projection development
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of ERK5 cascade
protein binding
receptor signaling protein tyrosine kinase activator activity
receptor tyrosine kinase binding
positive regulation of neuron projection development
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of ERK5 cascade
protein binding
receptor signaling protein tyrosine kinase activator activity
receptor tyrosine kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000344414.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401489.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000401503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000403610.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000405290.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000452023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000463919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | ENST00000619265.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200011450.hg.1 | NM_001002919.2 | Refseq |
