Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000379960.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381291.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000419241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000440794.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
NM_001001132.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
NM_003024.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
TC2100006976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381318.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000379960.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000381291.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399338.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399353.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000399367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000419241.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
ENST00000440794.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
NM_001001132.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100146934.hg.1 |
NM_003024.2 |
Refseq |