Assay Details
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005715.2 | NP_005706.1 | 4 - 5 | 823 | 146 | |
XM_011535378.2 | XP_011533680.1 | 4 - 5 | 1514 | 146 | ||
GenBank mRNA | AB020316.1 | - | 4 - 5 | 630 | 146 | |
AK292922.1 | - | 4 - 5 | 571 | 146 | ||
AK315320.1 | - | 4 - 5 | 531 | 146 | ||
BC093668.1 | - | 4 - 5 | 575 | 7939513 | 146 | |
BC093694.1 | - | 4 - 5 | 575 | 7939539 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001077.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
