Assay Details
Gene Symbol: PGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018290.3 | NP_060760.2 | 11 - 12 | 1483 | 152 | |
XM_011513711.2 | XP_011512013.1 | 12 - 13 | 1586 | 152 | ||
GenBank mRNA | AF109360.1 | - | 11 - 12 | 1423 | 152 | |
AK001845.1 | - | 11 - 12 | 1426 | 152 | ||
AK297752.1 | - | 9 - 10 | 1220 | 152 | ||
AL136705.1 | - | 11 - 12 | 1440 | 152 | ||
BC010087.2 | - | 11 - 12 | 1423 | 2964390 | 152 | |
CR457274.1 | - | 11 - 12 | 1410 | 152 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000213.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003153.hg.1 | ENST00000381967.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003153.hg.1 | ENST00000537241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04003153.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04003153.hg.1 | TC04000213.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000505986.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000510084.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000515668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | ENST00000625312.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007243.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | ENST00000512556.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | TC0400007243.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | NM_018290.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | ENST00000381967.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400149496.hg.1 | ENST00000512556.1 | Ensembl |
