Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
ENST00000247668.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
ENST00000359662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
ENST00000432785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
ENST00000536468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010835.hg.1 |
TC09000826.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
ENST00000247668.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
ENST00000359662.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
ENST00000432785.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
ENST00000482854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
ENST00000536468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010834.hg.1 |
TC09000826.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
TC0900009242.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163154.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163155.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163155.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900163155.hg.1 |
TC0900009242.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900163155.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900163155.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000247668.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000414589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000419057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000429509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000466107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000469701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000474950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
ENST00000482854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009242.hg.1 |
NM_021138.3 |
Refseq |