Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
ENST00000331772.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
ENST00000371947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
ENST00000466945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018906.hg.1 |
TC10001495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
ENST00000481793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011363.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
NM_018363.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
TC1000011363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000437752.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000331772.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000371947.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
ENST00000466945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
NM_001031709.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177203.hg.1 |
NM_018363.3 |
Refseq |