Assay Details
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011518973.2 | XP_011517275.1 | 1 - 2 | 261 | 134 | |
XM_011518976.2 | XP_011517278.1 | 1 - 2 | 261 | 103 | ||
GenBank mRNA | AK289722.1 | - | 1 - 2 | 104 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK289722.1, XM_011518976.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
regulation of immunoglobulin production
positive regulation of T cell cytokine production
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
I-kappaB kinase/NF-kappaB signaling
activation of NF-kappaB-inducing kinase activity
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
protein catabolic process
positive regulation of interleukin-2 production
tumor necrosis factor-mediated signaling pathway
negative regulation of glial cell apoptotic process
cellular protein-containing complex assembly
response to endoplasmic reticulum stress
regulation of apoptotic process
positive regulation of JUN kinase activity
positive regulation of T cell activation
positive regulation of DNA-binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
protein autoubiquitination
protein-containing complex assembly
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
protein homotrimerization
protein K63-linked ubiquitination
death-inducing signaling complex assembly
cellular response to nitric oxide
positive regulation of protein homodimerization activity
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
programmed necrotic cell death
negative regulation of neuron death
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of I-kappaB phosphorylation
positive regulation of extrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
CD40 receptor binding
protein binding
zinc ion binding
ligase activity
enzyme binding
protein kinase binding
protein phosphatase binding
mitogen-activated protein kinase kinase kinase binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
protein-containing complex binding
sphingolipid binding
regulation of immunoglobulin production
positive regulation of T cell cytokine production
activation of cysteine-type endopeptidase activity involved in apoptotic process
signal transduction
I-kappaB kinase/NF-kappaB signaling
activation of NF-kappaB-inducing kinase activity
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
protein catabolic process
positive regulation of interleukin-2 production
tumor necrosis factor-mediated signaling pathway
negative regulation of glial cell apoptotic process
cellular protein-containing complex assembly
response to endoplasmic reticulum stress
regulation of apoptotic process
positive regulation of JUN kinase activity
positive regulation of T cell activation
positive regulation of DNA-binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein heterooligomerization
protein autoubiquitination
protein-containing complex assembly
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
protein homotrimerization
protein K63-linked ubiquitination
death-inducing signaling complex assembly
cellular response to nitric oxide
positive regulation of protein homodimerization activity
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
programmed necrotic cell death
negative regulation of neuron death
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of I-kappaB phosphorylation
positive regulation of extrinsic apoptotic signaling pathway
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
CD40 receptor binding
protein binding
zinc ion binding
ligase activity
enzyme binding
protein kinase binding
protein phosphatase binding
mitogen-activated protein kinase kinase kinase binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
protein-containing complex binding
sphingolipid binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000247668.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000414589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000419057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000429509.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000466107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000469701.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000474950.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | ENST00000482854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009242.hg.1 | NM_021138.3 | Refseq |
