Assay Details
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011518976.2 | XP_011517278.1 | 2 - 3 | 299 | 121 | |
GenBank mRNA | AK289722.1 | - | 2 - 3 | 142 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK289722.1, XM_011518976.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
