Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
ENST00000342868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009749.hg.1 |
NM_197975.2 |
Refseq |