Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL834428.1 | - | 2 - 3 | 149 | 100 | |
BC010990.2 | - | 4 - 5 | 560 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
