Assay Details
Gene Symbol: BAG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017011284.1 | XP_016866773.1 | 12 - 13 | 2862 | 72 | |
XM_017011287.1 | XP_016866776.1 | 12 - 13 | 2835 | 72 | ||
XM_017011288.1 | XP_016866777.1 | 11 - 12 | 1625 | 72 | ||
XM_017011297.1 | XP_016866786.1 | 11 - 12 | 1625 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
