Assay Details
Gene Symbol: NDRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006723837.3 | XP_006723900.1 | 16 - 17 | 1067 | 65 | |
XM_006723838.3 | XP_006723901.1 | 14 - 15 | 1169 | 65 | ||
XM_011528925.2 | XP_011527227.1 | 14 - 15 | 979 | 65 | ||
XM_011528927.2 | XP_011527229.1 | 15 - 16 | 1031 | 65 | ||
XM_011528928.2 | XP_011527230.1 | 14 - 15 | 886 | 65 | ||
XM_017027978.1 | XP_016883467.1 | 16 - 17 | 1364 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
