Assay Details
Gene Symbol: TRAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011518973.2 | XP_011517275.1 | 2 - 3 | 329 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
